missing translation for 'onlineSavingsMsg'
Learn More

Silver AA Standard, Certified, 1000ppm Ag ±10ppm (1mL = 1mg Ag), LabChem™

Supplier:  LabChem LC224207

 View more versions of this product

Catalog No. LC224207


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver AA Standard
Amber Glass
99.84,0.16
1g/mL
AgNO3
AgNO3
Colorless
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
125 mL
CHEBI:32130
Certified
1000ppm (1mL = 1mg Ag)
7761-88-8,7732-18-5
Passes Test
Liquid
1000 ppm Ag ±10 ppm (1mL = 1mg Ag)
MFCD00003414
Soluble in water
[Ag+].[O-][N+]([O-])=O
169.87
24470
169.87

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
AgNO3
MFCD00003414
24470
silver(1+) nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions