missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver AA Standard, Certified, 1000ppm Ag ±10ppm (1mL = 1mg Ag), LabChem™
Supplier: LabChem LC224201
Specifications
| Silver AA Standard | |
| Amber Glass | |
| 99.84,0.16 | |
| 1g/mL | |
| AgNO3 | |
| AgNO3 | |
| Colorless | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 500 mL | |
| CHEBI:32130 | |
| Certified |
| 1000ppm (1mL = 1mg Ag) | |
| 7761-88-8,7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm Ag ±10 ppm (1mL = 1mg Ag) | |
| MFCD00003414 | |
| Soluble in water | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| 24470 | |
| 169.87 |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
| AgNO3 | |
| MFCD00003414 | |
| 24470 | |
| silver(1+) nitrate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature