missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicone Fluid, 1000, Spectrum™ Chemical
SI114, 63148-62-9
$256.52 - $23,449.45
Chemical Identifiers
| CAS | 63148-62-9 |
|---|---|
| Molecular Formula | (C2H6OSi)n |
| Molecular Weight (g/mol) | NaN |
| MDL Number | MFCD00132673 |
| IUPAC Name | Polydimethylsiloxane |
| SMILES | C[Si](C)(-*)O-* |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18606220
|
N/A | 100 g | Amber Glass Bottle |
Each for $256.52
|
|
||||
|
18606222
|
N/A | 44 lb. | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18606223
|
N/A | 440 lb. | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Silicone Fluid, 1000
Chemical Identifiers
| 63148-62-9 | |
| NaN | |
| Polydimethylsiloxane |
| (C2H6OSi)n | |
| MFCD00132673 | |
| C[Si](C)(-*)O-* |
Specifications
| 63148-62-9 | |
| (C2H6OSi)n | |
| MFCD00132673 | |
| Polydimethylsiloxane | |
| Ungraded |
| 1 | |
| 950-1050 cSt | |
| C[Si](C)(-*)O-* | |
| NaN | |
| Silicone Fluid, 1000 |