missing translation for 'onlineSavingsMsg'
Learn More

Silicone Fluid, 1000, Spectrum™ Chemical

Catalog No. 18606220
Change view
Click to view available options
Quantity:
100 g
44 lb.
440 lb.
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606220 100 g Amber Glass Bottle
18606222 44 lb. Amber Glass Bottle
18606223 440 lb. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606220 Supplier Spectrum Chemical Mfg Cor Supplier No. SI114100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

SI114, 63148-62-9

Spectrum™ Chemical Silicone Fluid, 1000

Chemical Identifiers

CAS 63148-62-9
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00132673
IUPAC Name Polydimethylsiloxane
SMILES C[Si](C)(-*)O-*

Specifications

CAS Max % 1
Quantity 100 g
Packaging Amber Glass Bottle
Viscosity 950-1050 cSt
Grade Ungraded
Chemical Name or Material Silicone Fluid, 1000