missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$309.71 - $542.00
Specifications
| Chemical Name or Material | Silicon Standard |
|---|---|
| Packaging | Triple-leached LDPE bottle |
| CAS | 16919-19-0 , 7732-18-5 |
| Certificate of Analysis | Assurance Certificate of Analysis reports up to 39 elements at ppb levels. |
| CAS Min % | 0.1,99.9 |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Si | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Silicon Standard | |
| 16919-19-0 , 7732-18-5 | |
| 0.1,99.9 | |
| 0.1,99.9 | |
| AA, ICP | |
| ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
| [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F | |
| diazanium;hexafluorosilicon(2-) | |
| 28145 | |
| Matrix Concentration Cat. No.H2O/tr. HF 1000μg/mL PLSI9-2Y, -2T, -2X 10,000 PLSI9-3Y, -3XH2O 1000μg/mL PLSI9A-2X 10,000 PLSI9A-3X |
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| F6H8N2Si | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Certified Reference Material | |
| H2O/0.4% F | |
| 178.153 | |
| Assurance |
Product intended for laboratory use only