missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon Standard, 1000mg/L in H2O/0.24 F (Assurance Grade), SPEX CertiPrep™
$141.46 - $141.46
Chemical Identifiers
| CAS | 16919-19-0 |
|---|---|
| Molecular Formula | F6H8N2Si |
| Molecular Weight (g/mol) | 178.153 |
| InChI Key | ITHIMUMYFVCXSL-UHFFFAOYSA-P |
| PubChem CID | 28145 |
| IUPAC Name | diazanium;hexafluorosilicon(2-) |
| SMILES | [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Si | 1000μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Chemical Identifiers
| 16919-19-0 | |
| 178.153 | |
| 28145 | |
| [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
| F6H8N2Si | |
| ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
| diazanium;hexafluorosilicon(2-) |
Specifications
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,99.9 | |
| AA, ICP | |
| Certified Reference Material | |
| [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F | |
| diazanium;hexafluorosilicon(2-) | |
| 30 mL | |
| Assurance | |
| Silicon Standard |
| 16919-19-0 , 7732-18-5 | |
| 0.1,99.9 | |
| F6H8N2Si | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
| Water | |
| 178.153 | |
| 28145 | |
| Matrix Concentration Cat. No.H2O/tr. HF 1000μg/mL PLSI9-2Y, -2T, -2X 10,000 PLSI9- |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient