missing translation for 'onlineSavingsMsg'
Learn More

Silicon ICP Standard, 1000 ppm Si in H2O/tr HF, Ricca Chemical

$329.32 - $622.83

Chemical Identifiers

CAS 16919-19-0
Molecular Formula F6H8N2Si
Molecular Weight (g/mol) 178.153
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PSI1KW-100
View Documents
Ricca Chemical Company
PSI1KW100
100 mL Natural LDPE Bottle
Each for $329.32
Only null left
 
PSI1KW-500
View Documents
Ricca Chemical Company
PSI1KW500
500 mL Natural Poly Bottle
Each for $622.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16919-19-0
178.153
28145
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
F6H8N2Si
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)

Specifications

Natural LDPE Bottle
97.0,0.62,0.01
100.0,0.64,0.01
Liquid
Colorless
Standard
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
178.153
28145
16919-19-0 , 7664-39-3 , 7732-18-5
1,000ppm Si
1 mL = 1 mg Si
F6H8N2Si
100°C
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)
100 mL
Silicon ICP Standard
Videos
SDS
Documents

Documents