missing translation for 'onlineSavingsMsg'
Learn More

Silicon ICP Standard, 10,000 ppm Si in H2O/tr HF, Ricca Chemical

Catalog No. PSI10KW100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PSI10KW100 100 mL Natural LDPE Bottle
PSI10KW500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PSI10KW100 Supplier Ricca Chemical Company Supplier No. PSI10KW100

Chemical Identifiers

CAS 16919-19-0 , 7664-39-3 , 7732-18-5
Molecular Formula F6H8N2Si
Molecular Weight (g/mol) 178.153
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Si
CAS Min % 92.0, 5.84, 0.09
CAS Max % 96.0, 6.08, 0.1
Equivalence 1 mL = 10 mg Si
Physical Form Liquid
Color Colorless
Type Standard
Quantity 100 mL
Chemical Name or Material Silicon ICP Standard