missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon ICP-MS Standard, 1000 ppm Si in H2O/tr HF, Ricca Chemical
Supplier: Ricca Chemical Company MSSI1KW100
Specifications
| Silicon ICP-MS Standard | |
| 16919-19-0 , 7664-39-3 , 7664-39-3 , 7732-18-5 | |
| 97.0,0.62,0.01 | |
| 1 mL = 1 mg Si | |
| F6H8N2Si | |
| 0.0°C | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
| diazanium;hexafluorosilicon(2-) | |
| 100 mL | |
| Laboratory |
| Natural LDPE Bottle | |
| 1,000ppm Si | |
| 100.0,0.64,0.01 | |
| Liquid | |
| Colorless | |
| 100°C | |
| Miscible with water | |
| ICP Single-element Standard Solution | |
| [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F | |
| 178.153 | |
| 28145 |
Chemical Identifiers
| 16919-19-0 | |
| 178.153 | |
| 28145 | |
| [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
| F6H8N2Si | |
| ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
| diazanium;hexafluorosilicon(2-) |