missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon AA Standard, 1000 ppm Si in H2O, Ricca Chemical
$107.00 - $318.40
Chemical Identifiers
| CAS | 6834-92-0 |
|---|---|
| Molecular Formula | Na2O3Si |
| Molecular Weight (g/mol) | 122.062 |
| InChI Key | NTHWMYGWWRZVTN-UHFFFAOYSA-N |
| PubChem CID | 23266 |
| ChEBI | CHEBI:60720 |
| IUPAC Name | disodium;dioxido(oxo)silane |
| SMILES | [O-][Si](=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
ASI1KW-100
|
Ricca Chemical Company
ASI1KW100 |
100 mL | Natural LDPE Bottle |
Each for $107.00
|
|
||||
|
ASI1KW-500
|
Ricca Chemical Company
ASI1KW500 |
500 mL | Natural Poly Bottle |
Each for $318.40
|
|
||||
Chemical Identifiers
| 6834-92-0 | |
| 122.062 | |
| 23266 | |
| disodium;dioxido(oxo)silane |
| Na2O3Si | |
| NTHWMYGWWRZVTN-UHFFFAOYSA-N | |
| CHEBI:60720 | |
| [O-][Si](=O)[O-].[Na+].[Na+] |
Specifications
| Natural LDPE Bottle | |
| 97.0,0.81 | |
| 100.0,0.84 | |
| Liquid | |
| Colorless | |
| NTHWMYGWWRZVTN-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)silane | |
| 100 mL | |
| CHEBI:60720 |