missing translation for 'onlineSavingsMsg'
Learn More

Silicon AA Standard, 1000 ppm Si in H2O, Ricca Chemical

Catalog No. ASI1KW100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ASI1KW100 100 mL Natural LDPE Bottle
ASI1KW500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ASI1KW100 Supplier Ricca Chemical Company Supplier No. ASI1KW100

Chemical Identifiers

CAS 6834-92-0 , 7732-18-5
Molecular Formula Na2O3Si
Molecular Weight (g/mol) 122.062
InChI Key NTHWMYGWWRZVTN-UHFFFAOYSA-N
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Si
CAS Min % 97.0, 0.81
CAS Max % 100.0, 0.84
Equivalence 1 mL = 1 mg Si
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 100 mL
Chemical Name or Material Silicon AA Standard