missing translation for 'onlineSavingsMsg'
Learn More

Silica Standard, 100 ppm SiO2 (46.7 ppm Si), Ricca Chemical

Catalog No. p-7239407
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
674516 500 mL Natural Poly Bottle
R67450001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 674516 Supplier Ricca Chemical Company Supplier No. 674516

Chemical Identifiers

CAS 6834-92-0 , 7732-18-5
Molecular Formula Na2O3Si
Molecular Weight (g/mol) 122.062
InChI Key NTHWMYGWWRZVTN-UHFFFAOYSA-N
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Silica Standard, 1 mL = 0.1 mg SiO2