missing translation for 'onlineSavingsMsg'
Learn More

Silica Standard, 100 ppm SiO2 (46.7 ppm Si), Ricca Chemical

Catalog No. R67450001A
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R67450001A 1 L Natural Poly Bottle
674516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R67450001A Supplier Ricca Chemical Company Supplier No. R67450001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6834-92-0 , 7732-18-5
Molecular Formula Na2O3Si
Molecular Weight (g/mol) 122.062
InChI Key NTHWMYGWWRZVTN-UHFFFAOYSA-N
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 100.0 ± 0.5ppm
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solubility Infinitely soluble in water
Solvent or Matrix Water
Quantity 1 L
Chemical Name or Material Silica Standard, 1mL = 0.1mg SiO2, 100ppm SiO2 (46.7ppm Si)