missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Standard, 10 ppm SiO2 (4.67 ppm Si), Ricca Chemical
$147.80 - $147.80
Chemical Identifiers
| CAS | 6834-92-0 |
|---|---|
| Molecular Formula | Na2O3Si |
| Molecular Weight (g/mol) | 122.062 |
| InChI Key | NTHWMYGWWRZVTN-UHFFFAOYSA-N |
| PubChem CID | 23266 |
| ChEBI | CHEBI:60720 |
| IUPAC Name | disodium;dioxido(oxo)silane |
| SMILES | [O-][Si](=O)[O-].[Na+].[Na+] |
Chemical Identifiers
| 6834-92-0 | |
| 122.062 | |
| 23266 | |
| disodium;dioxido(oxo)silane |
| Na2O3Si | |
| NTHWMYGWWRZVTN-UHFFFAOYSA-N | |
| CHEBI:60720 | |
| [O-][Si](=O)[O-].[Na+].[Na+] |
Specifications
| Natural Poly Bottle | |
| 98.0,0.01 | |
| 1 mL = 0.01 mg SiO | |
| Na2O3Si | |
| 100°C | |
| [O-][Si](=O)[O-].[Na+].[Na+] | |
| 122.062 | |
| 23266 | |
| Silica Standard |