missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel Desiccant, Indicating, 6 to 18 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$18290.67 - $18290.67
Chemical Identifiers
| CAS | 1343-98-2 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| PubChem CID | 14942 |
| ChEBI | CHEBI:26675 |
| IUPAC Name | silanedione |
| SMILES | O=[Si]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
2003901
|
Avantor J.T.Baker
340105 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 1343-98-2 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| CHEBI:26675 | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| 14942 | |
| silanedione |
Specifications
| 1343-98-2 | |
| 2.5 kg | |
| MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| Poly Bottle | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:26675 | |
| BAKER ANALYZED™ ACS Reagent |
| Silica Gel | |
| O2Si | |
| 6 to 18 Mesh | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| 14942 | |
| 1L = 2.10kg | |
| Silica Gel Desiccant, Indicating |