missing translation for 'onlineSavingsMsg'
Learn More

Sildenafil Citrate 98.0+%, TCI America™

Supplier:  TCI America S09861G

 View more versions of this product

Catalog No. S09861G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sildenafil Citrate
192°C
C28H38N6O11S
1 g
DEIYFTQMQPDXOT-UHFFFAOYSA-N
5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one;2-hydroxypropane-1,2,3-tricarboxylic acid
62853
666.70
Crystalline Powder
171599-83-0
White
MFCD09026931
sildenafil citrate, revatio, viagra, sildenafil citrate usan, sildenafilcitrate, unii-bw9b0ze037, caverta, sildenafil citrate salt, revatio tn
CCCC1=NN(C2=C1NC(=NC2=O)C3=C(C=CC(=C3)S(=O)(=O)N4CCN(CC4)C)OCC)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
666.703
CHEBI:58987
≥98.0% (HPLC)

Chemical Identifiers

171599-83-0
666.703
DEIYFTQMQPDXOT-UHFFFAOYSA-N
62853
5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one;2-hydroxypropane-1,2,3-tricarboxylic acid
C28H38N6O11S
MFCD09026931
sildenafil citrate, revatio, viagra, sildenafil citrate usan, sildenafilcitrate, unii-bw9b0ze037, caverta, sildenafil citrate salt, revatio tn
CHEBI:58987
CCCC1=NN(C2=C1NC(=NC2=O)C3=C(C=CC(=C3)S(=O)(=O)N4CCN(CC4)C)OCC)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TL4284390

TSCA : No

SDS
Documents

Documents

Promotions

Promotions