missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sildenafil Citrate 98.0+%, TCI America™
Supplier: TCI America S0986100MG
Specifications
| Sildenafil Citrate | |
| 192°C | |
| C28H38N6O11S | |
| 100 mg | |
| DEIYFTQMQPDXOT-UHFFFAOYSA-N | |
| 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one;2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 62853 | |
| 666.70 | |
| Crystalline Powder |
| 171599-83-0 | |
| White | |
| MFCD09026931 | |
| sildenafil citrate, revatio, viagra, sildenafil citrate usan, sildenafilcitrate, unii-bw9b0ze037, caverta, sildenafil citrate salt, revatio tn | |
| CCCC1=NN(C2=C1NC(=NC2=O)C3=C(C=CC(=C3)S(=O)(=O)N4CCN(CC4)C)OCC)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O | |
| 666.703 | |
| CHEBI:58987 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 171599-83-0 | |
| 666.703 | |
| DEIYFTQMQPDXOT-UHFFFAOYSA-N | |
| 62853 | |
| 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one;2-hydroxypropane-1,2,3-tricarboxylic acid |
| C28H38N6O11S | |
| MFCD09026931 | |
| sildenafil citrate, revatio, viagra, sildenafil citrate usan, sildenafilcitrate, unii-bw9b0ze037, caverta, sildenafil citrate salt, revatio tn | |
| CHEBI:58987 | |
| CCCC1=NN(C2=C1NC(=NC2=O)C3=C(C=CC(=C3)S(=O)(=O)N4CCN(CC4)C)OCC)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O |
Safety and Handling
RTECSNumber : TL4284390
TSCA : No