missing translation for 'onlineSavingsMsg'
Learn More

Sildenafil Citrate 98.0+%, TCI America™

Catalog No. S098625MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S098625MG 25 mg
S0986100MG 100 mg
S09861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S098625MG Supplier TCI America Supplier No. S098625MG
Only null left

Chemical Identifiers

CAS 171599-83-0
Molecular Formula C28H38N6O11S
Molecular Weight (g/mol) 666.703
MDL Number MFCD09026931
InChI Key DEIYFTQMQPDXOT-UHFFFAOYSA-N
Synonym sildenafil citrate, revatio, viagra, sildenafil citrate usan, sildenafilcitrate, unii-bw9b0ze037, caverta, sildenafil citrate salt, revatio tn
PubChem CID 62853
ChEBI CHEBI:58987
IUPAC Name 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCCC1=NN(C2=C1NC(=NC2=O)C3=C(C=CC(=C3)S(=O)(=O)N4CCN(CC4)C)OCC)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Melting Point 192°C
Color White
Quantity 25 mg
Formula Weight 666.70
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sildenafil Citrate
RTECSNumber TL4284390
TSCA No