missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SGK (SGK1), GST Tag Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Invitrogen™ PV3818

 View more versions of this product

Catalog No. PV3818


Only null left
Explore available promotions

Description

Description

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

SGK (SGK1) is a serine/threonine kinase that may be a transcriptional target for p38/MAPK in hepatoma cells. SGK plays an important role in cellular stress response and cell survival. SGK1 is transcriptionally activated by numerous stimuli, including serum, glucocorticoids, osmotic stress, and hormones. It phosphorylates and negatively regulates pro-apoptotic FOXO3A. SGK1 is thought to play an essential function in nephropathy and cardiovascular development.
Specifications

Specifications

O00141
Kinase Assay
6446
Human SGK (SGK1), GST Tag
Purified
-80°C, Avoid Freeze/Thaw Cycles
serine/threonine protein kinase SGK; serine/threonine-protein kinase Sgk1; serum and glucocorticoid-dependent protein kinase; serum/glucocorticoid regulated kinase 1; serum/glucocorticoid-regulated kinase 1; Sgk; SGK1; Sgk1 variant i3
SGK1
60-431
Unconjugated
Recombinant
60-431
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Dry Ice
SGK
Insect cells
Recombinant, Catalytic
Serine/Threonine Kinases
See Label
50mM tris with 0.02% Triton X-100, 0.5mM EDTA, 150mM NaCl, 2mM DTT, 50% glycerol and no preservative; pH 7.5
69.7 kDa
7.5
10 μg
RUO
SGK1
SGK Kinase Family
Protein
Human
GST-tag
-80°C, Avoid Freeze/Thaw Cycles
Ser⁄Thr Kinases: AGC Group
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
serum⁄glucocorticoid regulated kinase 1
Kinases & Inhibitors
Liquid
Signal Transduction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions