missing translation for 'onlineSavingsMsg'
Learn More

SGE Syringes, Cone Tip, Sold by MilliporeSigma™ Supelco™

Catalog No. 111010677
Change view
Click to view available options
Volume (Metric):
0.5 μL
5 μL
Needle Gauge:
23 G
26 G
Product Type:
Gastight™ Syringe
Liquid Syringe
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Needle Gauge Product Type
111010677 5 μL 23 G Gastight™ Syringe
111010678 5 μL 26 G Gastight™ Syringe
111010300 0.5 μL 23 G Liquid Syringe
111010695 5 μL 23 G Liquid Syringe
111010694 5 μL 26 G Liquid Syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111010677 Supplier Merck Emd Millipore Supplier No. 21929U

Designed and tested to meet critical autosampler specifications. For use with the PerkinElmer™ Autosystem.

  • Designed with a unique plunger to eliminate bending and seizing
  • Needle point style withstands multiple and fast septum injections
  • Stainless steel cone tip, point style #1 is recommended for use with pre-drilled septa
  • Needle shape on the syringe has been developed for multiple, fast septum injections on the autosampler
  • One of the most trouble-free autosampler syringes available

Specifications

Material Stainless Steel (Tip)
Color Lime
Needle Gauge 23 G
Product Type Gastight™ Syringe
Model 5 F-PE-GT-0.63
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 5 μL
Length (Metric) Needle 70 mm
Needle Point Style Cone
For Use With (Equipment) PerkinElmer Autosystem
Connection Type Fixed
Show More Show Less

PerkinElmer is a registered trademark of PerkinElmer Inc.