Learn More
MilliporeSigma™ Supelco™ SGE Syringes, Cone Tip
For use with Agilent/HP 7673, 7683 and 6850 ALS Autosamplers
Supplier: MilliporeSigma™ Supelco™ 26889U
Description
- Designed and tested to meet critical autosampler specifications
- A unique plunger eliminates bending and seizing
- Needle point style withstands multiple and fast septum injections
- One of the most trouble free autosampler syringes available
Specifications
| Stainless Steel (Tip) | |
| SGE Syringe | |
| 1 | |
| 10 μL | |
| Cone | |
| Cemented |
| 23 to 26 G | |
| 10 F-HP-0.63/0.47 | |
| 6 Pk. | |
| 42 mm | |
| Agilent 7673, 7683, 6850 ALS Autosampler |