Learn More
MilliporeSigma™ Supelco™ SGE MicroVolume Syringes, Standard Plunger
Equipped with bevel tip needles, designed for optimum septum penetration and to prevent septum coring
$74.09 - $128.39
Specifications
| Material | Stainless Steel (Tip) |
|---|---|
| Description | Standard Plunger |
| Needle Gauge | 25 G |
| Product Type | SGE Microvolume Syringe |
| Needle Point Number | 2 |
Description
- Sharp and clear fused graduation
- Extended graduation lines and wide white backing for reduced errors
- Angled needle tip for optimum septum penetration
- Standard, stainless steel plunger is individually fitted to its barrel
- Extended barrel guides the plunger during injection
- Suitable for training and student use
- Robust
Specifications
| Stainless Steel (Tip) | |
| 25 G | |
| 2 | |
| 50 mm | |
| Fixed |
| Standard Plunger | |
| SGE Microvolume Syringe | |
| 1 Ea. | |
| Bevel |