missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SGE Microvolume Syringe

A sample as small as 0.1 μL is contained in the needle

Supplier:  MilliporeSigma™ Supelco™ 509221

 View more versions of this product

Catalog No. 111010804


Only null left

Description

Description

  • Nozzle tip with 0.12 mm opening and cone shape for better septum
  • Full sample displacement
  • Plunger extended to the tip of the needle - Zero Dead Volume
  • No sample carryover
  • Improved sample volume reproducibility
Specifications

Specifications

Stainless Steel (Tip)
23 G
0.63 mm
1 BR-5
1 Ea.
50 mm
Plunger-in-needle Syringe
8 mm
SGE Microvolume Syringe
1
1 μL
Cone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions