Learn More
SGE Gastight™ Syringe, Fixed Needle (LTN), Sold by MilliporeSigma™ Supelco™
Suitable for manual dispenser/diluter applications. Maximum inertness for liquid or gas samples.
Supplier:  Merck Emd Millipore SGMA26261
 View more versions of this product
Catalog No. 111011235
Only null left
Description
Description
Specifications
Specifications
Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
0.50 mm
500 F-GT
1 Ea.
50 mm
Kel-F Luer Cones, Fixed
25 G
SGE Gas Tight Syringe
2
500 μL
Bevel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
GASTIGHT is a registered trademark of Hamilton Co.