Learn More
MilliporeSigma™ Supelco™ SGE Gas Tight Syringes, Luer Lock
Can be effectively used for the introduction of samples to the gas sampling and injection system (GASIS) inlet
Supplier: MilliporeSigma™ Supelco™ 509507
Description
- Maximum inertness for liquid or gas samples
- Suitable for manual disperser/diluter applications
- High accuracy of dispensed volume
- Wide variety of terminations
- Less time consuming
- Improved work flow analysis
Specifications
| Autoclavable | |
| SGE Gas Tight Syringe | |
| 1 Ea. | |
| Luer Lock, Kel-F Luer Cones |
| Borosilicate Glass (Barrel), PTFE (Plunger) | |
| 10 MDR-LL-GT | |
| 10 mL |