Learn More
Trajan™ BPX50 GC Capillary Columns
MS premium, low bleed phase designed for a range of EPA and pharmaceutical methods
$1653.51 - $1653.51
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity | |||
|
SG054741
|
Trajan™
054741 |
30 m | 0.15 mm | 0.15 μm |
Each for $1,653.51
|
|
|||
Description
- Mid-polarity column
- Inert
- Low bleed
- High temperature
- Ideal for a range of EPA methods and pharmaceutical applications
Specifications
| GC Capillary Column | |
| 350°C | |
| Mid polar |