missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Description
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
- Digitally controlled positive displacement dispensing system
- Programmable—enables user-independent liquid handling procedures
- Easily calibrated to ensure accurate dispensing
- Stores calibration factors for up to ten XCHANGE™ syringes
- Touch Wheel user interface
- Full color screen
Applications:
- Preparation of calibration standards
- Addition of internal standards
- Precise dispensing of non-aqueous liquids
- Sample dilution
Specifications
Specifications
| Needle Gauge | 25 G |
| Diameter (English) Inner Needle | 0.012 in. |
| Diameter (Metric) Inner Needle | 0.32 mm |
| Diameter (English) Outer Needle | 0.024 in. |
| Diameter (Metric) Outer Needle | 0.63 mm |
| For Use With (Application) | Preparation of calibration standards, addition of internal standards |
| Length (English) Needle | 1.96 in. |
| Product Type | eVol Analytical Syringe |
| Quantity | Ea. |
| Volume (Metric) | 500 μL |
| Show More |