missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SGE Autosampler Syringes, Cemented Needle (N)

Catalog No. 111010900
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
Needle Gauge:
23 G
26 G
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Needle Gauge
111010900 5 μL 26 G
111011378 10 μL 23 G
111011370 10 μL 26 G
111010618 25 μL 23 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111010900 Supplier MilliporeSigma™ Supelco™ Supplier No. 29623U

For use with CTC/LEAP PAL GC

Specifications

Material Stainless Steel (Tip)
Needle Gauge 26 G
Diameter (Metric) Outer Needle 0.47 mm
Product Type SGE Autosampler Syringe
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 5 μL
Length (Metric) Needle 50 mm
Needle Point Style Cone
For Use With (Equipment) CTC, LEAP
Connection Type Cemented
Show More Show Less