missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SGE Autosampler Syringes, Cemented Needle (N)
For use with CTC/LEAP PAL GC
Supplier: MilliporeSigma™ Supelco™ 29629U
Specifications
| Glass (Barrel), Stainless Steel (Tip) | |
| 23 G | |
| SGE Autosampler Syringe | |
| 1 Ea. | |
| 50 mm | |
| CTC, LEAP |
| Green | |
| 0.63 mm | |
| 1 | |
| 25 μL | |
| Cone | |
| Cemented |