Learn More
Trajan™ Autosampler Syringes
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
$198.42 - $215.89
| Catalog Number | Mfr. No. | Volume (Metric) | Needle Gauge | Connection Type | For Use With (Equipment) | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Needle Gauge | Connection Type | For Use With (Equipment) | Price | Quantity | ||
|
SG001982
|
Trajan™
001982 |
5 μL | 26 G | Fixed | Carlo Erba/Fisons/Leap/CTC AS200 and AS800, GC autosampler syringe |
Each for $215.89
|
|
||
|
SG002993
|
Trajan™
002993 |
10 μL | 26 G | Removable | Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus |
Each for $198.42
|
|
||
Description
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
| Autosampler Syringe | |
| Ea. |