Learn More
Trajan™ Autosampler Syringe Replacement Plungers for Thermoseparations/Spectraphysics
Accurate and reproducible injections. For use with AS100/300 & AS1000/3000/3500 LC Autosamplers
Supplier: Trajan™ SG006660
Description
- High quality solutions for liquid handling
- Specifically designed for Thermo Scientific instruments
Spécifications
| PTFE (Plunger) | |
| High performance liquid chromatology | |
| 250C-THERMOSPEC | |
| 250 μL |
| Gas tight | |
| Autosampler Syringe | |
| Ea. | |
| Thermo AS100/300 and AS1000/3000/3500 LC Autosamplers |