missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SGC GAK 1

Catalog No. 65645
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65645 5 mg
656425 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65645 Supplier Tocris Bioscience™ Supplier No. 6564/5*
Only null left

High affinity cyclin G associated kinase (GAK) inhibitor

SGC GAK 1 is a high affinity cyclin G associated kinase (GAK) inhibitor (Kd = 1.9 nM). Exhibits >30-fold selectivity for GAK over a panel of other kinases. Inhibits GAK in live cell assays (IC50 = 110 nM).

Specifications

Chemical Name or Material 6-Bromo-N-(3,4,5-trimethoxyphenyl)-4-quinolinamine
CAS 2226517-76-4
Quantity 5 mg
Molecular Formula C18H17BrN2O3
Purity 99%