missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ Seven2Go™ Pro pH/mV and Ion Meter: S8
Ideal for mobile applications in the laboratory, at-line, and outdoors
Manufacturer: Mettler Toledo™ 30207878
Includes: Instrument, LabX direct pH, USB cable, electrode clip, InLab Routine Pro ISM (includes cable), buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), bottles for calibration, uGo carrying case, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
Description
The Seven2Go portable meters are simple, reliable and are engineered to provide users with fast quality data, one-handed operation and durability.- New intuitive menu guarantees out-of-the-box operation for anyone
- T-Pad provides comfort and improves the speed with which you move through menus
- Smart button arrangement for faster navigation
- Automatically recognizes InLab ISM™ (Intelligent Sensor Management) sensor and stores sensor ID, serial number, and last calibration data, thereby reducing chances of error
- IP67 protection and drop test resistance make these meters weatherproof (withstand harsh and demanding environments)
- Supports 12 languages and offers a user interface that uses complete words and sentences
- Lack of cryptic expressions simplify operation and reduces strain on the user
- Sunlight-proof: direct sunlight enhances screen's contrast and guarantees an excellent readability even under bright and sunny conditions
- StatusLight™, an easy to understand traffic light helps to check if your meter is ready or not
- 5-Point calibration (pH and Ion)
- Linear and segmented calibration methods (pH and Ion)
- Graphic LCD display
- Ability to perform automatic, manual and time-dependent endpoint recording
- Perform serial measurements in user-defined time interval
- Acoustic and visual endpoint signal
- Other features include: Time and date option, three user levels (Routine, Expert, Outdoor), Calibration reminder, Sensor ID, SN, Sample ID, User ID, User-defined alarm limits, PIN protection
- Includes eight predefined buffer groups and one user-defined Ion-standards
- User-defined buffer group
- Automatic buffer recognition
- Automatic/Manual temperature capture
Specifications:
- pH Measuring Range: -2 to 20
- pH Resolution: 0.001/0.01/0.1
- pH Accuracy: ±0.002
- ORP/Redox Measuring Range: -2000 to +2000mV
- ORP/Redox Resolution: 0.1/1
- ORP/Redox Accuracy: ±0.1
- Units: mV, rel. mV
- Ion Measuring range: 1.00e-09 to 9.99e+09
- Ion Resolution: 3 or 4 digits (auto-range)
- Ion Accuracy: ±0.5%
- Units: mg/L, mmol/L, mol/L, %, ppm, pX
- Temperature Measuring Range: -5° to +130°C (ATC); -30° to +130°C (MTC)
- Temperature Resolution: 0.1°C
- Temperature Accuracy: ±0.2°C if T: 0° to 105°C; ±0.5°C if T<0°C or T>105°C
- Memory Size: 2000 (GLP conform)
- Sensor Inputs: BNC (1012 Ohm)/RCA (cinch)/2mm banana
- Communication interfaces: Micro-USB (USB Device)
- Power Supply: Four 1.5 V AA, 1.3 V NiMH acc. or USB
- Operating Hours: 200 to 250 hours
- Dimensions: 222 x 70 x 35mm
- Weight: 290g
Applications:
- S8-Standard kit: Wide range of samples, also suited for emulsions and suspensions.
- S8-Field kit: Wide range of samples, also suited for emulsions and suspensions; ideal for measurements on the go.
- S8-Biotech kit: Fast results in wide range of samples including harsh chemicals.
- S8-Fluoride kit: Aqueous samples, tooth paste and mouth wash.
Kits Available:
- Cat. No. 01-913-981, S8-Meter: Instrument, LabX direct pH, USB cable, electrode clip, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-982, S8-Standard kit: Instrument, LabX direct pH, USB cable, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-983, S8-Field kit: Instrument, LabX direct pH, USB cable, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), bottles for calibration, uGo™ carrying case, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-984, S8-Biotech kit: Instrument, LabX direct pH, USB cable, electrode clip, InLab™ Routine Pro ISM (includes cable), buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), bottles for calibration, uGo™ carrying case, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-985, S8-Fluoride kit: Instrument, LabX direct pH, USB cable, electrode clip, perfectION Fluoride, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
Certifications
ISO 9001, ISO 14001
Specifications
±0.5% | |
±0.002 | |
4 x AA or 1.3V NiMH acc. | |
ISO 9001, ISO 14001 | |
2000 (GLP conform) | |
Fast results in wide range of samples including harsh chemicals. | |
BNC (1012 Ohm)/RCA (cinch)/2mm banana | |
0.1, 1 | |
0.1°C | |
LCD | |
-2000 to 2000mV | |
0.001/0.01/0.1 |
±0.1 | |
200 to 250 hr. | |
5 | |
Micro-USB (USB Device) | |
222 x 70 x 35mm | |
Instrument, LabX direct pH, USB cable, electrode clip, InLab Routine Pro ISM (includes cable), buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), bottles for calibration, uGo carrying case, base accessory, operating instructions, quick guide, test report, | |
Seven2Go Pro S8 pH/mV and Ion Meter: Biotech Kit | |
±0.2°C if T: 0° to 105°C; ±0.5°C if T<0°C or T>105°C | |
290 g | |
IP 67 | |
-2 to 20 | |
-5° to +130°C (ATC); -30° to +130°C (MTC) |
Promotions
-
Earn Up to $2,000 in Accessories with Purchase
Buy METTLER TOLEDO balances or analytical instruments, you’ll get up to $2,000 to spend on accessories.: 06/30/21
-
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
Bundle your purchase of qualifying METTLER TOLEDO balances or analytical instruments to earn more.: 06/30/21