missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ Seven2Go™ pH/mV Meter: S2
Ideal for mobile applications in the laboratory, at-line, and outdoors
Manufacturer: Mettler Toledo™ 30207951
Description
The Seven2Go portable meters are simple, reliable and are engineered to provide users with fast quality data, one-handed operation and durability.- New intuitive menu guarantees out-of-the-box operation for anyone
- T-Pad provides comfort and improves the speed with which you move through menus
- Smart button arrangement for faster navigation
- Automatically recognizes InLab ISM™ (Intelligent Sensor Management) sensor and stores sensor ID, serial number, and last calibration data, thereby reducing chances of error
- IP67 protection and drop test resistance make these meters weatherproof (withstand harsh and demanding environments)
- 5-Point calibration (pH)
- Segmented LCD display
- Four predefined buffer groups
- Automatic buffer recognition
- Linear and segmented calibration methods
- With user-defined buffer group
- Ability to perform manual and time-dependent endpoint recording
- Perform serial measurements in user-defined time interval
- Visual endpoint signal
- Automatic/Manual temperature capture
Specifications:
- pH Measuring Range: -2 to 20
- pH Resolution: 0.01/0.1
- pH Accuracy: ±0.01
- ORP/Redox Measuring Range: -1999 to +1999mV
- ORP/Redox Resolution: 1
- ORP/Redox Accuracy: ±1
- Units: mV, rel. mV
- Temperature Measuring Range: -5° to +105°C
- Temperature Resolution: 0.1°C
- Temperature Accuracy: ±0.5°C
- ISM: Light version
- Memory Size: 200
- Sensor Inputs: BNC (1012 Ohm)/RCA (cinch)
- Power Supply: Four 1.5 V AA or 1.3 V NiMH acc.
- Operating Hours: 250 to 400 hours
- Dimensions: 222 x 70 x 35mm
- Weight: 270g
Applications:
- S2-Standard kit: Wide range of samples including emulsions and suspensions.
- S2-Field kit: Wide range of samples including emulsions and suspensions; ideal for measurements on the go.
- S2-Food kit: Solid and semi-solid samples like cheese and meat.
- S2-Light kit: Academic and educational purposes.
Kits Available:
- Cat. No. 01-913-965, S2-Meter: Instrument, electrode clip, operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-966, S2-Standard kit: Instrument, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-967, S2-Field kit: Instrument, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), bottles for calibration, uGo™ carrying case, operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-968, S2-Food kit: Instrument, electrode clip, InLab™ Solids Pro-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), bottles for calibration, uGo™ carrying case, operating instructions, quick guide, test report, declaration of conformity and batteries.
- Cat. No. 01-913-969, S2-Light kit: Instrument, electrode clip, InLab™ Versatile Pro, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), operating instructions, quick guide, test report, declaration of conformity and batteries.
Certifications
ISO 9001, ISO 14001
Specifications
±0.5°C | |
±0.01 | |
200 (Memory) | |
-5°C to +105°C | |
4 x AA or 1.3V NiMH acc. | |
222 x 70 x 35 mm | |
Instrument, pH electrode (InLab Expert Pro-ISM-IP67), electrode clip, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), bottles for calibration, uGo™ carrying case, operating instructions, quick guide, test report, declaration of conformity | |
IP-67 | |
-2 to 20 | |
1 | |
Seven2Go S2 pH/mV Meter: Field Kit w/ pH probe |
±1 | |
5 | |
LCD | |
250 to 400 hr. | |
ISO 9001, ISO 14001 | |
Fast results in aqueous samples; ideal for measurements on the go. | |
BNC (1012 Ohm)/RCA (cinch) (sensor) | |
-1999 to 1999 mV | |
0.01 to 0.1 | |
0.1°C | |
270 g |
Promotions
-
Earn Up to $2,000 in Accessories with Purchase
Buy METTLER TOLEDO balances or analytical instruments, you’ll get up to $2,000 to spend on accessories.: 06/30/21
-
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
Bundle your purchase of qualifying METTLER TOLEDO balances or analytical instruments to earn more.: 06/30/21