missing translation for 'onlineSavingsMsg'
Learn More

Sertraline Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135913
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05071G 1 g
S05075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05071G Supplier TCI America Supplier No. S05071G

Chemical Identifiers

CAS 79559-97-0
Molecular Formula C17H18Cl3N
Molecular Weight (g/mol) 342.688
MDL Number MFCD00895772
InChI Key BLFQGGGGFNSJKA-XHXSRVRCSA-N
Synonym (1S,4S)-1-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-4-(methylamino)naphthalene Hydrochloride
PubChem CID 63009
ChEBI CHEBI:9124
IUPAC Name (1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
SMILES CNC1CCC(C2=CC=CC=C12)C3=CC(=C(C=C3)Cl)Cl.Cl

Specifications

Physical Form Powder/Crystals
Color White
Melting Point 245°C
Quantity 1 g
Formula Weight 342.69
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Sertraline Hydrochloride
RTECSNumber QJ0352070
TSCA No