Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Series S Sensor Chip Protein A
Sensor chip for orientation-specific binding of antibodies through Fc-region
Supplier: Cytiva (Formerly GE Healthcare Life Sciences) 29127556
Description
- MabSelect™ SuRe ligand on the surface
- Binds only to heavy chain within the Fc region and ensures antibodies are bound to surface in specific orientation
- Ready-to-use sensor chip eliminates need to develop immobilization and regeneration conditions
- Highly controlled development processes and manufacturing conditions ensure reproducible results
- Carboxymethylated dextran matrix with a recombinant Protein A variant covalently attached
- Surface ligand same as used in MabSelect™ SuRe protein purification products
- Binds antibodies from several mammalian species (like human antibodies of the subclasses IgG1, IgG2, and IgG4)
- Binds only to heavy chain within the Fc region, ensuring specific orientation of antibody on the surface
- Does not bind Fab fragments (unlike native Protein A)
Specifications
| Surface carboxymethylated dextran pre-immobilized with recombinant Protein A (MabSelect™ SuRe ligand) | |
| Biacore 8 K+, Biacore 8 K, Biacore T200, Biacore S200, Biacore 4000 SPR Systems | |
| For Antibody Concentration and Interaction Analysis Using Biacore Systems |
| 3 x Sensor Chips | |
| 3 Pack |