missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Select

Catalog No. 09928107
Change view
Click to view available options
Quantity:
5 mL
60 mL
450 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09928107 60 mL
09928106 5 mL
09928108 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09928107 Supplier Cytiva Supplier No. 29343052
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Delivers selective binding of DNA fragments for use with next-generation sequencing (NGS) and PCR clean-up.

Sera-Mag Select PCR clean-up and size selection reagent combines the exceptional binding characteristics of Sera-Mag Carboxyl Speedbeads with an optimized binding solution in a ready-to-use formulation. The DNA size range isolated from the process can be tailored to suit the user’s end requirements by adjusting the amount of reagent that is mixed with a fixed volume of sample.

  • Use a single reagent for PCR clean-up and size selection
  • Easily control what size DNA is retained or eliminated by adding various ratios of Sera-Mag Select to your sample, enabling NGS library and PCR fragment clean-up, as well as dual size selection of libraries
  • Uses the same protocols as existing reagents so it can be directly integrated into existing workflows with minimal disruption
  • Sera-Mag Select delivers tight distribution in size selection for high yield recovery of specific fragment sizes
  • Suitable for manual, high throughput and automated workflows
  • Sera-Mag Select should be stored in the refrigerator between 2°C to 8°C - Do not freeze - Store upright and keep bottle tightly sealed

Specifications

Content And Storage 2°C to 8°C
Isolation Technology Magnetic Bead
For Use With (Application) Tested functionally in size selection
Product Type Size Selection and PCR Clean-Up Reagent
Quantity 60 mL