Learn More
Applied Biosystems™ Sequence Detection Primer

Sequence Detection Primer
$69.50 - $207.00
Specifications
| For Use With (Equipment) | 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System |
|---|---|
| Product Type | Sequencing Primer |
| Content And Storage | 1 tube containing an unlabeled primer in solution or dried down. Store at -15°C to -25°C |
| Purification Method | Desalted |
| Shipping Condition | Room Temperature |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
4304970
![]()
|
Applied Biosystems™
4304970 |
10,000 pmol |
Each for $69.50
|
|
|||||
4304971
![]()
|
Applied Biosystems™
4304971 |
80,000 pmol |
Each for $114.00
|
|
|||||
4304972
![]()
|
Applied Biosystems™
4304972 |
130,000 pmol |
Each for $207.00
|
|
|||||
Description
These unlabeled primers can be used with TaqMan™ probes in any real-time PCR application. Choose between dry and in-solution formatted delivery. All primers are desalted.
Sequence detection primers are unlabeled primers that can be used with TaqMan probes or SYBR Green dye for your real-time PCR research applications.
Features include:
- Choice of delivered scales
- For use in any real-time PCR or PCR application
- All sequence detection primers are desalted
Real-time PCR sequence detection primers are delivered in either dry or liquid format.
Guaranteed yield
The unit size you select is based on the minimum amount we will deliver, not on synthesis starting amount.
Approximate ship time
4–6 days in North America, 5-10 days in Europe
We also offer:
- Custom TaqMan probes with a variety of options for dyes and quenchers.
- TaqMan MGB probes and primers available as predesigned and pre-formulated TaqMan Assays.
- Commercial manufacturing products and services, including OEM and GMP oligo manufacturing.
Need a different dye or concentration or a larger lot size? The TaqMan Custom Assay and Oligo Service offers custom products that you can tailor to meet your experimental needs.
Specifications
| 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System | |
| 1 tube containing an unlabeled primer in solution or dried down. Store at -15°C to -25°C |
|
| Room Temperature | |
| Applied Biosystems | |
| Liquid or Dried Down |
| Sequencing Primer | |
| Desalted | |
| Unlabeled | |
| Real Time PCR Applications |
For Research Use Only. Not for use in diagnostic procedures.