missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequence Detection Primer
GREENER_CHOICE

Catalog No. 4304972
Encompass
Change view
Click to view available options
Quantity:
10,000 pmol
130,000 pmol
80,000 pmol
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4304972 130,000 pmol
4304970 10,000 pmol
4304971 80,000 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4304972 Supplier Applied Biosystems™ Supplier No. 4304972
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sequence Detection Primer

These unlabeled primers can be used with TaqMan™ probes in any real-time PCR application. Choose between dry and in-solution formatted delivery. All primers are desalted.

Sequence detection primers are unlabeled primers that can be used with TaqMan probes or SYBR Green dye for your real-time PCR research applications.

Features include:

  • Choice of delivered scales
  • For use in any real-time PCR or PCR application
  • All sequence detection primers are desalted

Real-time PCR sequence detection primers are delivered in either dry or liquid format.

Guaranteed yield

The unit size you select is based on the minimum amount we will deliver, not on synthesis starting amount.

Approximate ship time

4–6 days in North America, 5-10 days in Europe

We also offer:

  • Custom TaqMan probes with a variety of options for dyes and quenchers.
  • TaqMan MGB probes and primers available as predesigned and pre-formulated TaqMan Assays.
  • Commercial manufacturing products and services, including OEM and GMP oligo manufacturing.

Need a different dye or concentration or a larger lot size? The TaqMan Custom Assay and Oligo Service offers custom products that you can tailor to meet your experimental needs.

Specifications

For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
Product Type Sequencing Primer
Content And Storage 1 tube containing an unlabeled primer in solution or dried down.

Store at -15°C to -25°C
Purification Method Desalted
Shipping Condition Room Temperature
Primer Unlabeled
Quantity 130,000 pmol
Product Line Applied Biosystems
For Use With (Application) Real Time PCR Applications
Form Liquid or Dried Down
I have purchased custom oligos (primers and probes) and I want to make a 20x working stock for a TaqMan assay, similar to the pre-designed assays. How do I dilute and mix my oligos to make a 20x working stock?

Most pre-designed TaqMan assays are supplied as a 20x assay concentrate containing 18 uM of each primer and 5 uM of probe which provides a 900 nM of each primer and 250 nM of the probe (for gene expression assays) and 200 nM of each probe (for SNP genotyping assays) at the final 1x concentration.

In order to make a 20x working stock from custom oligos, the oligos need to be diluted as follows: To make a 20x Assay stock for Gene Expression assays, make 100 uM stocks of both forward and reverse primers, and TaqMan probe. Add 18 ul of each primer and 5 ul of the TaqMan probe, which gives 41 ul volume. Add 59 ul of 1X TE to bring the volume to 100 ul of 20X TaqMan Gene Expression Assay. To make a 20x Assay stock for a Genotyping Assay, make 100 uM stocks of both forward and reverse primers, and both TaqMan probes. Add 18 ul of each primer and 4 ul of each TaqMan probe, which gives 44 ul volume. Add 56 ul of 1X TE to bring the volume to 100 ul of 20X TaqMan SNP Genotyping Assays.

What is the difference between Custom Standard DNA Oligos and Sequence Detection Primer (Cat. No. 4304970)?

The main difference between Custom Standard DNA Oligos and Sequence Detection Primer (Cat. Nos. 4304970, 4304971, 4304972) is that Sequence Detection Primers come in the guaranteed specific amount and concentration that is ordered. Whereas Custom DNA Oligos can be selected by the Synthesis Scale. The scale of synthesis (25 nmol, 50 nmol, 200 nmol, 1 µmol, or 10 µmol) refers to the starting scale and not the guaranteed final amount.

You can find more information about the Synthesis Scale by going to the Custom Standard DNA Oligos page and clicking on the question mark next to “Synthesis Scale.”

In addition, the sequence detection primers are desalted, whereas the custom DNA oligos can be purified by HPLC, PAGE, and Cartridge.

Which GAPDH primer set do you recommend for a SYBR Green dye-based real-time PCR assay?

We do not have pre-designed forward and reverse primers for GAPDH to be used in a SYBR Green dye-based real-time PCR assay. You can order either Sequence Detection Primers or Custom Standard DNA Oligos.

For Research Use Only. Not for use in diagnostic procedures.