missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sensor Chip CM4

Catalog No. 501055323
Change view
Click to view available options
Quantity:
3 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501055323 3 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501055323 Supplier Cytiva Supplier No. BR100539

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Biacore SPR systems. Use when sample contaminants have a high positive charge. Matrix: similar to Sensor Chipcm5 but with a lower degree of carboxymethylation, which can improve sensitivity for certain interactions. Due to the reduced net negative charge, the surface is well suited for use with negatively charged proteins and nucleic acids.
  • Use when sample contaminants have a high postive charge.
  • Improves sensitivity when monitoring certain interactions.
  • Suitable for negatively charged proteins and nucleic acids.
  • Attach proteins, nucleic acids, carbohydrates or small molecules.
  • Couple to carboxyl groups on the sensor surface via -NH&superscript_2;, -SH, -CHO, -OH or -COOH.

Specifications

Description Sensor Chip CM4
Kit Contents 3 x Sensor Chips
Matrix Carboxymethylated Dextran (with a lower degree of carboxymethylation compared to Sensor Chip CM5)
Quantity 3 Pack
For Use With (Application) For most types of interaction analyses in Biacore systems