Learn More
Sensidyne™ Detector Tubes: Nitrogen Compounds
Supplier:  Sensidyne™ 117SB
 View more versions of this product
Catalog No. 18072F
Only null left
Description
Description
All detector tubes are sold in packs of 10.
Specifications
Chemical Identifiers
10
Specifications
Nitrogen Dioxide
2
1.5 min.
0°C to 40°C
0.5 to 30ppm
10
32°F to 104°F
Product Suggestions
Product Suggestions