Learn More
Sensidyne™ Detector Tubes: Nitrogen Compounds
Supplier:  Sensidyne™ 233S
 View more versions of this product
Catalog No. 18072D
Only null left
Description
Description
All detector tubes are sold in packs of 10.
Specifications
Chemical Identifiers
10
Specifications
Nitric Acid Vapor
1
1.5 min.
5° to 40°C
1 to 20ppm
10
41°F to 104°F
Product Suggestions
Product Suggestions