missing translation for 'onlineSavingsMsg'
Learn More

Sensidyne™ Detector Tubes: Chlorine

Detector tubes

$187.58

Specifications

Components Measured Chlorine
Quantity 10
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 1
Catalog Number Mfr. No. Measurement Range Response Time No. of Strokes Price Quantity & Availability  
Catalog Number Mfr. No. Measurement Range Response Time No. of Strokes Price Quantity & Availability  
18068P
View Documents Promotion Details
Sensidyne™
109SB
0.1 to 10 1 min. 1/4/5
Pack of 10 Each for $187.58
Only null left
 
Description

Description

All detector tubes are sold in packs of 10.
Specifications

Specifications

Chlorine
10
32°F to 104°F
0°C to 40°C
Videos
SDS
Documents

Documents