missing translation for 'onlineSavingsMsg'
Learn More

Sensidyne™ Detector Tubes: Acetone

Detector tubes

$178.54

Specifications

Components Measured Acetone
No. of Strokes 1
Quantity 10
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 1
Catalog Number Mfr. No. Measurement Range Response Time Price Quantity & Availability  
Catalog Number Mfr. No. Measurement Range Response Time Price Quantity & Availability  
18066Q
View Documents Promotion Details
Sensidyne™
102SD
20 to 5000 1.5 min.
Pack of 10 Each for $178.54
Only null left
 
Description

Description

All detector tubes are sold in packs of 10.
Specifications

Specifications

Acetone
10
0°C to 40°C
1
32°F to 104°F
Videos
SDS
Documents

Documents