Learn More
Sensidyne™ Detector Tubes: Acetone
Catalog No. 18066Q
Change view
Click to view available options
Measurement Range:
20 to 5000
Response Time:
1.5 min.
Catalog No. Measurement Range Response Time
18066Q 20 to 5000 1.5 min.
1 options
Catalog No. 18066Q Supplier Sensidyne™ Supplier No. 102SD
Detector tubes
All detector tubes are sold in packs of 10.
Specifications
Components Measured Acetone
Measurement Range 20 to 5000
No. of Strokes 1
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C