Learn More
Ansell MICROFLEX™ Sensation™ N73 Nitrile Gloves
Powder free nitrile exam glove feature textured fingertips to provide a secure grip when working with small instruments.
$13.82 - $13.82
Specifications
Certifications/Compliance ISO9001 QMS, FDA21 CFR 177-2600, ASTM D6319
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Applications Healthcare, Laboratory
AQL 1.5
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-170-810
View Documents
Ansell
N730
X-Small
Pack of 100 Each for $13.82
Case of 10 Pack for $138.20
Only null left
19-170-830
View Documents
Ansell
N731
Small
Pack of 100 Each for $13.82
Case of 10 Pack for $138.20
Only null left
19-170-812
View Documents
Ansell
N732
Medium
Pack of 100 Each for $13.82
Case of 10 Pack for $138.20
Only null left
19-170-832
View Documents
Ansell
N733
Large
Pack of 100 Each for $13.82
Case of 10 Pack for $138.20
Only null left
19-170-814
View Documents
Ansell
N734
X-Large
Pack of 100 Each for $13.82
Case of 10 Pack for $138.20
Only null left
Description
Description
  • Thin, nitrile glove clinically proven to reduce redness and itching associated with contact dermatitis
  • Extra-soft with advanced tactile sensitivity and comfort
  • Enhanced comfort
  • Textured fingers
  • Very low sulfur and aluminum content
  • FDA cleared
  • Ideal for applications including assembly of parts, sample taking and processing and standard patient exams
Specifications
Specifications
ISO9001 QMS, FDA21 CFR 177-2600, ASTM D6319
No
1.5
Single-use
3.9 mil
Yes
2.4 mil
Powder Free
0.10 mm
Sensation™ N73
9.5 in.
EN 420:2003 + A1:2009
Healthcare, Laboratory
Blue
Partially Textured
0.10 mm
Nitrile
0.06 mm
3.9 mil
Yes
29 MPa
24.13 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'