missing translation for 'onlineSavingsMsg'
Learn More

Selenium AA Standard, 1000 ppm Se in H2O, Ricca Chemical

Catalog No. ASE1KW100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ASE1KW100 100 mL Natural LDPE Bottle
ASE1KW500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ASE1KW100 Supplier Ricca Chemical Company Supplier No. ASE1KW100

Chemical Identifiers

CAS 7446-08-4 , 7732-18-5
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
PubChem CID 24007
IUPAC Name (oxo-λ4-selanylidene)oxidane
SMILES O=[Se]=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Se
CAS Min % 98.0, 0.14
CAS Max % 100.0, 0.14
Equivalence 1 mL = 1 mg Se
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 100 mL
Chemical Name or Material Selenium AA Standard