missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Calcium ionophore II, Function Tested, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
$573.23 - $2137.30
Chemical Identifiers
| MDL Number | MFCD00043337 |
|---|---|
| Synonym | N,N,N′,N′-Tetra[cyclohexyl]diglycolic acid diamide; N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide; ETH 129 |
Chemical Identifiers
| MFCD00043337 | |
| N,N,N′,N′-Tetra[cyclohexyl]diglycolic acid diamide; N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide; ETH 129 |
Specifications
| 74267-27-9 | |
| C28H48N2O3 | |
| 50 mg | |
| 460.69 | |
| Selectophore |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD00043337 | |
| N,N,N′,N′-Tetra[cyclohexyl]diglycolic acid diamide; N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide; ETH 129 | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH