missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Calcium ionophore II, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111015400
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015400 250 mg
111015401 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015400 Supplier Merck Emd Millipore Supplier No. 21193250MGF

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00043337
Synonym N,N,N′,N′-Tetra[cyclohexyl]diglycolic acid diamide; N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide; ETH 129

Specifications

CAS 74267-27-9
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C28H48N2O3
Quantity 250 mg
Formula Weight 460.69
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH