missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Calcium ionophore II, Function Tested, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 74267-27-9 |
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| Linear Formula | C28H48N2O3 |
| Quantity | 50 mg |
| Formula Weight | 460.69 |
| Grade | Selectophore |
| Product Line | Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH